CAS RN 10605-09-1
CAS RN 10605-09-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
6-O-Stearoyl-L-ascorbic Acid 95.0+%, TCI America™
CAS: 10605-09-1 Molecular Formula: C24H42O7 Molecular Weight (g/mol): 442.593 MDL Number: MFCD00059739 InChI Key: LITUBCVUXPBCGA-WMZHIEFXSA-N Synonym: L-Ascorbyl 6-Stearate PubChem CID: 54725318 IUPAC Name: [(2S)-2-[(2R)-3,4-dihydroxy-5-oxo-2H-furan-2-yl]-2-hydroxyethyl] octadecanoate SMILES: CCCCCCCCCCCCCCCCCC(=O)OCC(C1C(=C(C(=O)O1)O)O)O