missing translation for 'onlineSavingsMsg'
Learn More

6-O-Stearoyl-L-ascorbic Acid 95.0+%, TCI America™

Catalog No. A06175G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A06175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A06175G Supplier TCI America Supplier No. A06175G
Only null left

Chemical Identifiers

CAS 10605-09-1
Molecular Formula C24H42O7
Molecular Weight (g/mol) 442.593
MDL Number MFCD00059739
InChI Key LITUBCVUXPBCGA-WMZHIEFXSA-N
Synonym L-Ascorbyl 6-Stearate
PubChem CID 54725318
IUPAC Name [(2S)-2-[(2R)-3,4-dihydroxy-5-oxo-2H-furan-2-yl]-2-hydroxyethyl] octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OCC(C1C(=C(C(=O)O1)O)O)O

Specifications

Melting Point 117°C
Color White-Yellow
Quantity 5 g
Formula Weight 442.59
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 6-O-Stearoyl-L-ascorbic Acid
EINECSNumber (5)-0063
RTECSNumber CI7671310
TSCA No