accessibility menu, dialog, popup

UserName

OOBr

CAS RN 10602-01-4

IUPAC Name: 2-(4-bromophenyl)-1,3-dioxolane
Synonyms: 2-(4-bromophenyl)-1,3-dioxolane
Molecular Weight (g/mol): 229.07
Molecular Formula: C9H9BrO2
InChi Key: ZYIMHOWVWWHLDN-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)C1OCCO1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)