missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Bromophenyl)-1,3-dioxolane 98.0+%, TCI America™

Catalog No. B21005G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21005G 5 g
B210025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B21005G Supplier TCI America Supplier No. B21005G

Chemical Identifiers

CAS 10602-01-4
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
MDL Number MFCD01318950
InChI Key ZYIMHOWVWWHLDN-UHFFFAOYSA-N
PubChem CID 2773842
IUPAC Name 2-(4-bromophenyl)-1,3-dioxolane
SMILES C1COC(O1)C2=CC=C(C=C2)Br

Specifications

Color White-Yellow
Quantity 5 g
Synonym 2-4-bromophenyl-1,3-dioxolane, 1-bromo-4-1,3-dioxolan-2-yl benzene, 4-bromobenzaldehyde ethylene acetal, 1,3-dioxolane,2-4-bromophenyl, 2-4'-bromophenyl-1,3-dioxolane, 1,3-dioxolane, 2-4-bromophenyl, bromophenyldioxolane, acmc-2098ib, 2-4-bromophenyl dioxolane, 4-bromobenzaldehyde ethylene ketal
Formula Weight 229.07
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-(4-Bromophenyl)-1,3-dioxolane