accessibility menu, dialog, popup

UserName

CAS RN 106-44-5

IUPAC Name: 4-methylphenol
Synonyms: P-cresol aluminum salt 4-cresol p-kresol p-toluol p-cresol for synthesis paramethyl phenol ccris 647 potassium salt 4-cresol mixed cresols sodium salt 4-cresol
Molecular Weight (g/mol): 108.14
Molecular Formula: C7H8O
InChi Key: IWDCLRJOBJJRNH-UHFFFAOYSA-N
SMILES: CC1=CC=C(O)C=C1
Boiling Point 
  • (4)
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (4)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (8)
  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)
  • (4)
  • (1)