missing translation for 'onlineSavingsMsg'
Learn More

p-Cresol 99.0+%, TCI America™

Catalog No. C0400500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0400500G 500 g
C040025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0400500G Supplier TCI America Supplier No. C0400500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 106-44-5
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
MDL Number MFCD00002376
InChI Key IWDCLRJOBJJRNH-UHFFFAOYSA-N
Synonym p-cresol, 4-cresol, 4-hydroxytoluene, p-methylphenol, phenol, 4-methyl, p-hydroxytoluene, p-tolyl alcohol, p-kresol, para-cresol, p-oxytoluene
PubChem CID 2879
ChEBI CHEBI:17847
IUPAC Name 4-methylphenol
SMILES CC1=CC=C(C=C1)O

Specifications

Color White-Yellow
Boiling Point 202°C
Quantity 500 g
UN Number 3455
Formula Weight 108.14
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material p-Cresol
EINECSNumber (3)-0499&(4)-0057
RTECSNumber GO6475000
TSCA Yes