missing translation for 'onlineSavingsMsg'
Learn More

p-Cresol, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020480
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020480 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020480 Supplier Merck Emd Millipore Supplier No. 424295GF

p-cresol is a volatile phenolic compound, having a tar like odor, which is a metabolite of tyrosine and phenylalanine, mainly produced by the intestinal bacteria.

Agriculture, Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

Ignition Point 1038°F
CAS 106-44-5
Vapor Pressure 1 mmHg (20°C)
Vapor Density 3.72 (vs. air)
Density 1.034 g/mL (at 25°C)
Flash Point 85°C
Physical Form Neat
Linear Formula CH3C6H4 OH
MDL Number MFCD00002376
Melting Point 32°C to 34°C; 32°C to 35°C
Boiling Point 202°C
Synonym 4-Methylphenol
Type Analytical Standard
Quantity 5 g
Formula Weight 108.14
Percent Purity ≥99.0% (GC)
Grade Analytical Standard
Show More Show Less