accessibility menu, dialog, popup

UserName

CAS RN 10513-45-8

IUPAC Name: 2-tert-butyl-5-methyl-2H-1λ⁴,2-oxazol-1-ylium perchlorate
Synonyms: 2-tert-butyl-5-methyl-1λ⁴,2-oxazol-1-ylium perchlorate
Molecular Weight (g/mol): 239.65
Molecular Formula: C8H14ClNO5
InChi Key: MPSXGPCFLAGJOM-UHFFFAOYSA-M
SMILES: [O-][Cl](=O)(=O)=O.CC1=[O+]N(C=C1)C(C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results