missing translation for 'onlineSavingsMsg'
Learn More

N-tert-Butyl-5-methylisoxazolium Perchlorate 98.0+%, TCI America™

Catalog No. B08325G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B08325G 5 g
B08321G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B08325G Supplier TCI America Supplier No. B08325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10513-45-8
Molecular Formula C8H14ClNO5
Molecular Weight (g/mol) 239.65
MDL Number MFCD00011960
InChI Key MPSXGPCFLAGJOM-UHFFFAOYSA-M
PubChem CID 151520
IUPAC Name 2-tert-butyl-5-methyl-2H-1λ⁴,2-oxazol-1-ylium perchlorate
SMILES [O-][Cl](=O)(=O)=O.CC1=[O+]N(C=C1)C(C)(C)C

Specifications

Melting Point 119°C
Quantity 5 g
UN Number 1479
Synonym Woodward′s Reagent L
Formula Weight 239.65
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-tert-Butyl-5-methylisoxazolium Perchlorate