accessibility menu, dialog, popup

UserName

CAS RN 10489-74-4

IUPAC Name: ethyl (N'-hydroxycarbamimidoyl)formate
Synonyms: ethyl (N'-hydroxycarbamimidoyl)formate
Molecular Weight (g/mol): 132.12
Molecular Formula: C4H8N2O3
InChi Key: QGYKRMZPOOILBA-UHFFFAOYSA-N
SMILES: CCOC(=O)C(N)=NO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results