missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-oximinooxamate, 98%, Thermo Scientific Chemicals

Catalog No. AC426880050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC426880050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC426880050 Supplier Thermo Scientific Chemicals Supplier No. 426880050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10489-74-4 | Molecular Formula: C4H8N2O3 | Molecular Weight: 132.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10489-74-4
Molecular Formula C4H8N2O3
Molecular Weight (g/mol) 132.12
MDL Number MFCD03013442 MFCD14666500 MFCD03013442
InChI Key QGYKRMZPOOILBA-UHFFFAOYSA-N
Synonym ethyl 2-oximinooxamate, ethyl 2-amino-2-hydroxyimino acetate, ethyl z-2-amino-2-hydroxyimino acetate, ethyl 2-hydroxyamino-2-iminoacetate, ethyl n'-hydroxycarbamimidoyl formate, ethyl aminohydroxyiminoacetate, acetic acid, 2-amino-2-hydroxyimino-, ethyl ester, 2z, ethyl amino hydroxyimino ethanoate, ethyl n-hydroxycarbamimidoyl formate, amino-hydroxyimino-acetic acid ethyl ester
PubChem CID 9614369
IUPAC Name ethyl (2Z)-2-amino-2-hydroxyiminoacetate
SMILES CCOC(=O)C(N)=NO

Specifications

Melting Point 99°C to 103°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula HONHC(=NH)COOCH2CH3
Quantity 5 g
Formula Weight 132.12
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 2-oximinooxamate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously wit

RUO – Research Use Only