accessibility menu, dialog, popup

UserName

CAS RN 10482-55-0

CH3H3COOCH3CH3(Z)

CAS RN 10482-55-0

IUPAC Name: 3-methylbutyl (2Z)-2-methylbut-2-enoate
Synonyms: 3-methylbutyl (2Z)-2-methylbut-2-enoate
Molecular Weight (g/mol): 170.25
Molecular Formula: C10H18O2
InChi Key: ZARFDQHJMNVNLE-UITAMQMPSA-N
SMILES: C\C=C(\C)C(=O)OCCC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results