missing translation for 'onlineSavingsMsg'
Learn More

Isoamyl Angelate 99.0+%, TCI America™

Catalog No. A113125ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A113125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A113125ML Supplier TCI America Supplier No. A113125ML
Only null left

Chemical Identifiers

CAS 10482-55-0
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.25
MDL Number MFCD00213789
InChI Key ZARFDQHJMNVNLE-UITAMQMPSA-N
Synonym Angelic Acid Isoamyl Ester, Isoamyl (Z)-2-Methyl-2-butenoate, (Z)-2-Methyl-2-butenoic Acid Isoamyl Ester
PubChem CID 5366155
IUPAC Name 3-methylbutyl (2Z)-2-methylbut-2-enoate
SMILES C\C=C(\C)C(=O)OCCC(C)C

Specifications

Color Colorless
Boiling Point 201°C
Quantity 25 mL
Formula Weight 170.25
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Isoamyl Angelate