accessibility menu, dialog, popup

UserName

CAS RN 1047684-56-9

H3CSNNSCH3

CAS RN 1047684-56-9

IUPAC Name: 4,4'-bis(5-hexylthiophen-2-yl)-2,2'-bipyridine
Synonyms: 4,4'-bis(5-hexylthiophen-2-yl)-2,2'-bipyridine
Molecular Weight (g/mol): 488.75
Molecular Formula: C30H36N2S2
InChi Key: DCFNCZSDXIPGOQ-UHFFFAOYSA-N
SMILES: CCCCCCC1=CC=C(S1)C1=CC(=NC=C1)C1=NC=CC(=C1)C1=CC=C(CCCCCC)S1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results