missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Bis(5-hexyl-2-thienyl)-2,2'-bipyridyl 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 200 mg |
| Formula Weight | 488.75 |
| Percent Purity | ≥98.0% (HPLC,N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4,4′-Bis(5-hexyl-2-thienyl)-2,2′-bipyridyl |
Safety and Handling
| TSCA | No |