accessibility menu, dialog, popup

UserName

NH3(R)H2N

CAS RN 103831-11-4

IUPAC Name: (3R)-3-azaniumylpyrrolidin-1-ium
Synonyms: (3R)-3-ammoniopyrrolidin-1-ium
Molecular Weight (g/mol): 88.15
Molecular Formula: C4H12N2
InChi Key: NGXSWUFDCSEIOO-SCSAIBSYSA-P
SMILES: [NH3+][C@@H]1CC[NH2+]C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)