accessibility menu, dialog, popup

UserName

CAS RN 103204-69-9

IUPAC Name: 4-acetamido-2-methylbenzoic acid
Synonyms: 4-acetamido-2-methylbenzoic acid
Molecular Weight (g/mol): 193.2
Molecular Formula: C10H11NO3
InChi Key: AQPDTYYKDYMCTH-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC(C)=C(C=C1)C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)