accessibility menu, dialog, popup

UserName

CAS RN 1031-15-8

IUPAC Name: methyltriphenylphosphanium chloride
Synonyms: methyltriphenylphosphanium chloride
Molecular Weight (g/mol): 312.78
Molecular Formula: C19H18ClP
InChi Key: QRPRIOOKPZSVFN-UHFFFAOYSA-M
SMILES: [Cl-].C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)