Learn More
Methyltriphenylphosphonium chloride, 97%, Thermo Scientific Chemicals
Description
Methyltriphenylphosphonium chloride is used as a phase transfer catalyst in synthetic chemistry. It acts as a catalyst for regio-controlled ring-opening polymerization of substituted epoxides. It is utilized for controlling the corrosion iron. It acts as a reactant for intramolecular nitrone cycloaddition. In addition to this, it is used for neoplasm inhibition through antimitochondrial effect.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyltriphenylphosphonium chloride is used as a phase transfer catalyst in synthetic chemistry. It acts as a catalyst for regio-controlled ring-opening polymerization of substituted epoxides. It is utilized for controlling the corrosion iron. It acts as a reactant for intramolecular nitrone cycloaddition. In addition to this, it is used for neoplasm inhibition through antimitochondrial effect.
Solubility
Soluble in methanol.
Notes
Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 221°C (decomposition) |
| Linear Formula | CH3P(C6H5)3Cl |
| Quantity | 5 g |
| UN Number | UN3077 |
| Sensitivity | Hygroscopic |
| Solubility Information | Soluble in methanol. |
| Formula Weight | 312.78 |
| Percent Purity | 97% |
| Chemical Name or Material | Methyltriphenylphosphonium chloride |
Safety and Handling
| Hazard Category | H302+H312-H315-H318 |
| Hazard Statement | GHS H Statement H318-H302-H312-H315 Causes serious eye damage. Harmful if swallowed. Harmful in contact with skin. Causes skin irritation. |
| Precautionary Statement | P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
| EINECSNumber | 418-400-2 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only