accessibility menu, dialog, popup

UserName

OOS

CAS RN 103-04-8

IUPAC Name: 2-(phenylsulfanyl)acetate
Synonyms: phenylthioglycolic acid
Molecular Weight (g/mol): 167.2
Molecular Formula: C8H7O2S
InChi Key: MOTOSAGBNXXRRE-UHFFFAOYSA-M
SMILES: [O-]C(=O)CSC1=CC=CC=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)