CAS RN 10294-34-5
Boron trichloride, 1M soln. in hexanes, mixed isomers
CAS: 10294-34-5 Molecular Formula: BCl3 Molecular Weight (g/mol): 117.16 MDL Number: MFCD00011313 InChI Key: FAQYAMRNWDIXMY-UHFFFAOYSA-N Synonym: boron trichloride,borane, trichloro,boron chloride,trichloroboron,borontrichloride,chlorure de bore french,unii-k748471rag,hsdb 326,boron trichloride un1741 poison gas PubChem CID: 25135 IUPAC Name: trichloroborane SMILES: ClB(Cl)Cl
Boron trichloride, 1M solution in methylene chloride, AcroSeal™
CAS: 10294-34-5 | BCl3 | 117.16 g/mol
Boron trichloride, 1M soln. in dichloromethane, stab.
CAS: 10294-34-5 Molecular Formula: BCl3 Molecular Weight (g/mol): 117.16 MDL Number: MFCD00011313 InChI Key: FAQYAMRNWDIXMY-UHFFFAOYSA-N Synonym: boron trichloride,borane, trichloro,boron chloride,trichloroboron,borontrichloride,chlorure de bore french,unii-k748471rag,hsdb 326,boron trichloride un1741 poison gas PubChem CID: 25135 IUPAC Name: trichloroborane SMILES: ClB(Cl)Cl
Boron trichloride, 1M solution in hexane, AcroSeal™
CAS: 10294-34-5 | BCl3 | 117.16 g/mol
Boron Trichloride (ca. 16% in Heptane, ca. 1.0mol/L), TCI America™
CAS: 10294-34-5 Molecular Formula: BCl3 Molecular Weight (g/mol): 117.16 MDL Number: MFCD00011313 InChI Key: FAQYAMRNWDIXMY-UHFFFAOYSA-N Synonym: boron trichloride,borane, trichloro,boron chloride,trichloroboron,borontrichloride,chlorure de bore french,unii-k748471rag,hsdb 326,boron trichloride un1741 poison gas PubChem CID: 25135 IUPAC Name: trichloroborane SMILES: ClB(Cl)Cl
Boron Trichloride (ca. 9% in Dichloromethane, ca. 1.0mol/L), TCI America™
CAS: 10294-34-5 Molecular Formula: BCl3 Molecular Weight (g/mol): 117.16 MDL Number: MFCD00011313 InChI Key: FAQYAMRNWDIXMY-UHFFFAOYSA-N Synonym: boron trichloride,borane, trichloro,boron chloride,trichloroboron,borontrichloride,chlorure de bore french,unii-k748471rag,hsdb 326,boron trichloride un1741 poison gas PubChem CID: 25135 IUPAC Name: trichloroborane SMILES: ClB(Cl)Cl
Boron Trichloride (ca. 13% in p-Xylene, ca. 1.0mol/L), TCI America™
CAS: 10294-34-5 Molecular Formula: BCl3 Molecular Weight (g/mol): 117.16 MDL Number: MFCD00011313 InChI Key: FAQYAMRNWDIXMY-UHFFFAOYSA-N Synonym: boron trichloride,borane, trichloro,boron chloride,trichloroboron,borontrichloride,chlorure de bore french,unii-k748471rag,hsdb 326,boron trichloride un1741 poison gas PubChem CID: 25135 IUPAC Name: trichloroborane SMILES: ClB(Cl)Cl
Boron Trichloride (ca. 13% in Toluene, ca. 1.0mol/L), TCI America™
CAS: 10294-34-5 Molecular Formula: BCl3 Molecular Weight (g/mol): 117.16 MDL Number: MFCD00011313 InChI Key: FAQYAMRNWDIXMY-UHFFFAOYSA-N Synonym: boron trichloride,borane, trichloro,boron chloride,trichloroboron,borontrichloride,chlorure de bore french,unii-k748471rag,hsdb 326,boron trichloride un1741 poison gas PubChem CID: 25135 IUPAC Name: trichloroborane SMILES: ClB(Cl)Cl
Boron Trichloride (ca. 17% in Hexane, ca. 1.0mol/L), TCI America™
CAS: 10294-34-5 Molecular Formula: BCl3 Molecular Weight (g/mol): 117.16 MDL Number: MFCD00011313 InChI Key: FAQYAMRNWDIXMY-UHFFFAOYSA-N Synonym: boron trichloride,borane, trichloro,boron chloride,trichloroboron,borontrichloride,chlorure de bore french,unii-k748471rag,hsdb 326,boron trichloride un1741 poison gas PubChem CID: 25135 IUPAC Name: trichloroborane SMILES: ClB(Cl)Cl