CAS RN 10242-01-0
CAS RN 10242-01-0
IUPAC Name:
5-methoxy-1H-indole-3-carboxylic acid
Synonyms:
5-methoxy-1H-indole-3-carboxylic acid
Molecular Weight (g/mol):
191.19
Molecular Formula:
C10H9NO3
InChi Key:
RVVSEZGJCOAUED-UHFFFAOYSA-N
SMILES:
COC1=CC=C2NC=C(C(O)=O)C2=C1
1
–
1
of
1
results
1
5-Methoxyindole-3-carboxylic Acid Hydrate 98.0+%, TCI America™
CAS: 10242-01-0 Molecular Formula: C10H9NO3 Molecular Weight (g/mol): 191.186 MDL Number: MFCD03265451 InChI Key: RVVSEZGJCOAUED-UHFFFAOYSA-N Synonym: 5-methoxyindole-3-carboxylic acid,5-methoxy-3-indolecarboxylic acid,5-methoxy-3-carboxyindole,1h-indole-3-carboxylic acid, 5-methoxy,pubchem7984,zlchem 1276,1h-indole-3-carboxylicacid, 5-methoxy,5-methoxyindole-3-carboxylic acid;,5-methoxyindole-3-carboxylic acid 5-methoxy-1h-indazole-3-carboxylic acid PubChem CID: 259191 IUPAC Name: 5-methoxy-1H-indole-3-carboxylic acid SMILES: COC1=CC2=C(C=C1)NC=C2C(=O)O