accessibility menu, dialog, popup

UserName

CAS RN 102392-83-6

IUPAC Name: pyrrolidine-1-carboximidamide hydroiodide
Synonyms: pyrrolidine-1-carboximidamide hydroiodide
Molecular Weight (g/mol): 241.08
Molecular Formula: C5H12IN3
InChi Key: XPASGQSDLPDWNV-UHFFFAOYSA-N
SMILES: I.NC(=N)N1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results