accessibility menu, dialog, popup

UserName

CAS RN 1023674-80-7

IUPAC Name: 9-(3-bromophenyl)-10-phenylanthracene
Synonyms: 9-(3-bromophenyl)-10-phenylanthracene
Molecular Weight (g/mol): 409.33
Molecular Formula: C26H17Br
InChi Key: KHPNWZVLPLJEHH-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(=C1)C1=C2C=CC=CC2=C(C2=CC=CC=C2)C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results