missing translation for 'onlineSavingsMsg'
Learn More

9-(3-Bromophenyl)-10-phenylanthracene 98.0+%, TCI America™

Catalog No. B48975G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48975G 5 g
B48971G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48975G Supplier TCI America Supplier No. B48975G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1023674-80-7
Molecular Formula C26H17Br
Molecular Weight (g/mol) 409.33
MDL Number MFCD28100940
InChI Key KHPNWZVLPLJEHH-UHFFFAOYSA-N
PubChem CID 59302132
IUPAC Name 9-(3-bromophenyl)-10-phenylanthracene
SMILES BrC1=CC=CC(=C1)C1=C2C=CC=CC2=C(C2=CC=CC=C2)C2=CC=CC=C12

Specifications

Melting Point 184°C
Color White-Yellow
Quantity 5 g
Formula Weight 409.33
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-(3-Bromophenyl)-10-phenylanthracene