accessibility menu, dialog, popup

UserName

CAS RN 10191-60-3

IUPAC Name: [bis(methylsulfanyl)methylidene](cyano)amine
Synonyms: [bis(methylsulfanyl)methylidene](cyano)amine
Molecular Weight (g/mol): 146.23
Molecular Formula: C4H6N2S2
InChi Key: IULFXBLVJIPESI-UHFFFAOYSA-N
SMILES: CSC(SC)=NC#N
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)