missing translation for 'onlineSavingsMsg'
Learn More

S,S'-Dimethyl N-Cyanodithioiminocarbonate 98.0+%, TCI America™

Catalog No. D179825G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D179825G 25 g
D1798250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D179825G Supplier TCI America Supplier No. D179825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10191-60-3
Molecular Formula C4H6N2S2
Molecular Weight (g/mol) 146.226
MDL Number MFCD00009825
InChI Key IULFXBLVJIPESI-UHFFFAOYSA-N
Synonym dimethyl cyanocarbonimidodithioate, dimethyl cyanodithioiminocarbonate, dimethyl n-cyanodithioiminocarbonate, carbonimidodithioic acid, cyano-, dimethyl ester, s,s-dimethyl cyanoimidodithiocarbonate, cyanoimino bis methylthio methane, dimethyl n-cyanoimidodithiocarbonate, dimethyl cyanimidodithiocarbonate, dimethyl cyanoiminodithiocarbonate, dimethyl n-cyanodithiocarbonimidate
PubChem CID 66289
IUPAC Name bis(methylsulfanyl)methylidenecyanamide
SMILES CSC(=NC#N)SC

Specifications

Melting Point 54°C
Color Yellow
Quantity 25 g
UN Number 1759
Formula Weight 146.23
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material S,S′-Dimethyl N-Cyanodithioiminocarbonate