CAS RN 101712-20-3
IUPAC Name:
1-(2,6-difluorophenyl)propan-2-one
Synonyms:
1-(2,6-difluorophenyl)propan-2-one
Molecular Weight (g/mol):
170.16
Molecular Formula:
C9H8F2O
InChi Key:
LAWHOFKPDMZDLJ-UHFFFAOYSA-N
SMILES:
CC(=O)CC1=C(F)C=CC=C1F
1
–
2
of
2
results
1
2,6-Difluorophenylacetone, 98%, Thermo Scientific™
CAS: 101712-20-3 Molecular Formula: C9H8F2O Molecular Weight (g/mol): 170.16 InChI Key: LAWHOFKPDMZDLJ-UHFFFAOYSA-N Synonym: 1-2,6-difluorophenyl propan-2-one,2-propanone, 1-2,6-difluorophenyl,2,6-difluorophenylacetone PubChem CID: 2734927 IUPAC Name: 1-(2,6-difluorophenyl)propan-2-one SMILES: CC(=O)CC1=C(C=CC=C1F)F
2
2,6-Difluorophenylacetone, 98%, Thermo Scientific Chemicals
CAS: 101712-20-3 Molecular Formula: C9H8F2O Molecular Weight (g/mol): 170.16 InChI Key: LAWHOFKPDMZDLJ-UHFFFAOYSA-N Synonym: 1-2,6-difluorophenyl propan-2-one,2-propanone, 1-2,6-difluorophenyl,2,6-difluorophenylacetone PubChem CID: 2734927 IUPAC Name: 1-(2,6-difluorophenyl)propan-2-one SMILES: CC(=O)CC1=C(C=CC=C1F)F