accessibility menu, dialog, popup

UserName

CAS RN 101711-78-8

IUPAC Name: (4S)-4-benzyl-3-propanoyl-1,3-oxazolidin-2-one
Synonyms: (4S)-4-benzyl-3-propanoyl-1,3-oxazolidin-2-one
Molecular Weight (g/mol): 233.27
Molecular Formula: C13H15NO3
InChi Key: WHOBYFHKONUTMW-NSHDSACASA-N
SMILES: CCC(=O)N1[C@@H](CC2=CC=CC=C2)COC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results