accessibility menu, dialog, popup

UserName

HN

CAS RN 1013-88-3

IUPAC Name: diphenylmethanimine
Synonyms: benzenemethanimine, α-phenyl-
Molecular Weight (g/mol): 181.24
Molecular Formula: C13H11N
InChi Key: SXZIXHOMFPUIRK-UHFFFAOYSA-N
SMILES: N=C(C1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (3)
  • (4)
  • (2)
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)
  • (5)