accessibility menu, dialog, popup

UserName

CAS RN 10114-58-6

IUPAC Name: dihydrogen 4-(2-{3-[2-(2,4-diaminophenyl)diazen-1-yl]phenyl}diazen-1-yl)benzene-1,3-diamine dichloride
Synonyms: dihydrogen bismarck brown Y dichloride
Molecular Weight (g/mol): 419.31
Molecular Formula: C18H20Cl2N8
InChi Key: MCZVRBLCRZWFJH-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].NC1=CC=C(N=NC2=CC(=CC=C2)N=NC2=CC=C(N)C=C2N)C(N)=C1
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results