missing translation for 'onlineSavingsMsg'
Learn More

Bismarck Brown, TCI America™

Catalog No. B078025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B078025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B078025G Supplier TCI America Supplier No. B078025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10114-58-6
Molecular Formula C18H20Cl2N8
Molecular Weight (g/mol) 419.314
MDL Number MFCD00012977
InChI Key MCZVRBLCRZWFJH-UHFFFAOYSA-N
Synonym basic brown 1, bismark brown y, bismarck brown y, c.i. basic brown 1, dihydrochloride, bismarck brown b, unii-14mub626s2, phenylene brown, manchester brown, 1,3-bis 2,3-diaminophenylazo benzene hydrochloride, 1,3-benzenediamine, 4,4'-1,3-phenylenebis azo bis-, dihydrochloride
PubChem CID 82360
ChEBI CHEBI:53615
IUPAC Name 4-[[3-[(2,4-diaminophenyl)diazenyl]phenyl]diazenyl]benzene-1,3-diamine;dihydrochloride
SMILES C1=CC(=CC(=C1)N=NC2=C(C=C(C=C2)N)N)N=NC3=C(C=C(C=C3)N)N.Cl.Cl

Specifications

Quantity 25 g
Formula Weight 419.31
Physical Form Crystalline Powder
Chemical Name or Material Bismarck Brown
EINECSNumber (5)-2037
TSCA Yes