accessibility menu, dialog, popup

UserName

CAS RN 101084-20-2

ONHBrH2N

CAS RN 101084-20-2

IUPAC Name: 3-amino-5-bromo-1,4-dihydropyridin-4-one
Synonyms: 3-amino-5-bromo-1H-pyridin-4-one
Molecular Weight (g/mol): 189.01
Molecular Formula: C5H5BrN2O
InChi Key: DMZVRJHJGBNINH-UHFFFAOYSA-N
SMILES: NC1=CNC=C(Br)C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results