missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-bromo-4-hydroxypyridine, 95%

Catalog No. AAH66810MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66810MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH66810MD Supplier Thermo Scientific Chemicals Supplier No. H66810MD
Only null left

CAS: 101084-20-2 | C5H5BrN2O | 189.012 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 101084-20-2
Molecular Formula C5H5BrN2O
Molecular Weight (g/mol) 189.012
MDL Number MFCD08692340
InChI Key DMZVRJHJGBNINH-UHFFFAOYSA-N
Synonym 3-amino-5-bromopyridin-4-ol, 3-amino-5-bromopyridin-4 1h-one, 3-amino-5-bromo-pyridin-4-ol, 4-pyridinol,3-amino-5-bromo, 4-pyridinol, 3-amino-5-bromo, 3-amino-5-bromo-4-hydroxypyridine
PubChem CID 21560634
IUPAC Name 3-amino-5-bromo-1H-pyridin-4-one
SMILES C1=C(C(=O)C(=CN1)Br)N

Specifications

Quantity 250 mg
Beilstein 121085
Formula Weight 189.01
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-Amino-5-bromo-4-hydroxypyridine
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only