missing translation for 'onlineSavingsMsg'
Learn More

Cadmium chloride, anhydrous, ACS, 99.0% min

Catalog No. AA3662936
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3662936 500 g
AA3662922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3662936 Supplier Thermo Scientific Chemicals Supplier No. 03662936

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10108-64-2 | CdCl2 | 183.31 g/mol

Cadmium chloride is a precursor material for the preparation of cadmium sulfide. It is mainly utilized in dyeing, pigments, photocopying and electroplating of cadmium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Preparation of cadmium sulfide, as analytical lab reagent, photography, manufacturing of special mirrors, ice-nucleating agent, lubricant, to absorb H2S in sulfide analysis

Chemical Identifiers

CAS 10108-64-2
Molecular Formula CdCl2
Molecular Weight (g/mol) 183.31
MDL Number MFCD00010916
InChI Key YKYOUMDCQGMQQO-UHFFFAOYSA-L
Synonym cadmium chloride, cadmium dichloride, caddy, vi-cad, kadmiumchlorid, caswell no. 135, ccris 114, hsdb 278, cadmium ii chloride
PubChem CID 24947
ChEBI CHEBI:35456
IUPAC Name dichlorocadmium
SMILES [Cl-].[Cl-].[Cd++]

Specifications

Boiling Point 960°C
Melting Point 568°C
Physical Form Powder
Quantity 500 g
Odor Odorless
Sensitivity Hygroscopic
Density 4.047 g/mL
UN Number UN2570
Merck Index 14,1617
Solubility Information Soluble in water,acetone; Slightly soluble in methanol; Insoluble in ether.
Formula Weight 183.32
Percent Purity ≤99.0%
Grade ACS
Chemical Name or Material Cadmium chloride, Anhydrous
Show More Show Less
Hazard Category H301-H330-H335-H340-H350-H360FD-H372
Hazard Statement GHS H Statement
H301-H330-H340-H350-H360-H372
Toxic if swallowed.
Fatal if inhaled.
May cause genetic defects.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P284-P301+P310-P304+P340-P308+P313-P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS
EINECSNumber 233-296-7
RTECSNumber EV0175000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only