missing translation for 'onlineSavingsMsg'
Learn More

Cadmium chloride, ACS reagent, anhydrous

Catalog No. AC315270050
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC315270050 5 g Glass bottle
AC315271000 100 g Plastic bottle
AC315275000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC315270050 Supplier Thermo Scientific Chemicals Supplier No. 315270050

missing translation for 'validMainframeMsg'

CAS: 10108-64-2 | CdCl2 | 183.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10108-64-2
Molecular Formula CdCl2
Molecular Weight (g/mol) 183.31
MDL Number MFCD00010916
InChI Key YKYOUMDCQGMQQO-UHFFFAOYSA-L
Synonym cadmium chloride, cadmium dichloride, caddy, vi-cad, kadmiumchlorid, caswell no. 135, ccris 114, hsdb 278, cadmium ii chloride
PubChem CID 24947
ChEBI CHEBI:35456
IUPAC Name dichlorocadmium
SMILES [Cl-].[Cl-].[Cd++]

Specifications

Boiling Point 960.0°C
Melting Point 568.0°C
Color White
Physical Form Powder
Quantity 5 g
Linear Formula CdCl2
Fieser 04,485
Merck Index 15,1622
Solubility Information Solubility in water: 1400g/L (20°C). Other solubilities: soluble in acetone, slightly soluble in methanol and ethanol, practically insoluble in ether
Formula Weight 183.31
Percent Purity ≥99.0%
Grade ACS Reagent
Packaging Glass bottle
Chemical Name or Material Cadmium chloride, Anhydrous, For analysis ACS
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause genetic defects.
Causes damage to organs through prolonged or repeated exposure.
Fatal if inhaled.
May cause cancer.
May damage fertility.
May damage the unborn child.
Toxic if swallowed
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Obtain special instructions before use.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF S
EINECSNumber 233-296-7

RUO – Research Use Only