accessibility menu, dialog, popup

UserName

CAS RN 101023-55-6

IUPAC Name: 1H-imidazole hydrobromide
Synonyms: imidazole hydrobromide
Molecular Weight (g/mol): 148.99
Molecular Formula: C3H5BrN2
InChi Key: VWUCIBOKNZGWLX-UHFFFAOYSA-N
SMILES: Br.N1C=CN=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)