accessibility menu, dialog, popup

UserName

H3CCH3OOHNCl

CAS RN 101-21-3

IUPAC Name: propan-2-yl N-(3-chlorophenyl)carbamate
Synonyms: chlorpropham chlorpropham (iso) 氯普芬
Molecular Weight (g/mol): 213.66
Molecular Formula: C10H12ClNO2
InChi Key: CWJSHJJYOPWUGX-UHFFFAOYSA-N
SMILES: CC(C)OC(=O)NC1=CC=CC(Cl)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results