missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Chlorpropham, MilliporeSigma™ Supelco™
Description
Agriculture and Environmental
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 101-21-3 |
| Physical Form | Neat |
| Linear Formula | C10H12 NO2 Cl |
| MDL Number | MFCD00037108 |
| Synonym | Chloro-ICP; Chloropropham; Isopropyl N-(3-chlorophenyl)carbamate |
| Type | Analytical Standard |
| Quantity | 250 mg |
| Formula Weight | 213.66 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH