CAS RN 101-20-2
IUPAC Name:
3-(4-chlorophenyl)-1-(3,4-dichlorophenyl)urea
Synonyms:
triclocarban cutisan TCC nobacter solubacter triclocarbanum 3,4,4'-trichlorodiphenylurea
Molecular Weight (g/mol):
315.58
Molecular Formula:
C13H9Cl3N2O
InChi Key:
ICUTUKXCWQYESQ-UHFFFAOYSA-N
SMILES:
ClC1=CC=C(NC(=O)NC2=CC=C(Cl)C(Cl)=C2)C=C1
1
–
3
of
3
results
3
3,4,4'-Trichlorocarbanilide 98.0+%, TCI America™
CAS: 101-20-2 Molecular Formula: C13H9Cl3N2O Molecular Weight (g/mol): 315.58 MDL Number: MFCD00013254 InChI Key: ICUTUKXCWQYESQ-UHFFFAOYSA-N PubChem CID: 7547 ChEBI: CHEBI:48347 IUPAC Name: 3-(4-chlorophenyl)-1-(3,4-dichlorophenyl)urea SMILES: ClC1=CC=C(NC(=O)NC2=CC=C(Cl)C(Cl)=C2)C=C1