accessibility menu, dialog, popup

UserName

CAS RN 10083-24-6

IUPAC Name: 5-[(1E)-2-(3,4-dihydroxyphenyl)ethenyl]benzene-1,3-diol
Synonyms: piceatannol 3-hydroxyresveratol 3,3',4'5-tetrahydroxystilbene 3,5,3',4'-tetrahydroxystilbene
Molecular Weight (g/mol): 244.25
Molecular Formula: C14H12O4
InChi Key: CDRPUGZCRXZLFL-OWOJBTEDSA-N
SMILES: OC1=CC(\C=C\C2=CC=C(O)C(O)=C2)=CC(O)=C1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)