missing translation for 'onlineSavingsMsg'
Learn More

Piceatannol, 96%, Thermo Scientific™

Catalog No. AC328450010
Change view
Click to view available options
1mg; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC328450010 1mg; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC328450010 Supplier Thermo Scientific Chemicals Supplier No. 328450010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 10083-24-6
Infrared Spectrum Authentic
Assay Percent Range 96%
Molecular Formula C14H12O4
MDL Number MFCD00221715
Quantity 1 mg
Synonym piceatannol, 3-hydroxyresveratol, astringinin, piceatanol, e-4-3,5-dihydroxystyryl benzene-1,2-diol, 3,3',4'5-tetrahydroxystilbene, 3,5,3',4'-tetrahydroxystilbene, 4-e-2-3,5-dihydroxyphenyl ethenyl benzene-1,2-diol, 3,3',4,5'-tetrahydroxystilbene, unii-6ks3ls0d4f
InChI Key CDRPUGZCRXZLFL-OWOJBTEDSA-N
SMILES C1=CC(=C(C=C1C=CC2=CC(=CC(=C2)O)O)O)O
IUPAC Name 4-[(E)-2-(3,5-dihydroxyphenyl)ethenyl]benzene-1,2-diol
Molecular Weight (g/mol) 244.246
PubChem CID 667639
ChEBI CHEBI:28814
Formula Weight 244.3
Percent Purity ≥95%
Chemical Name or Material Piceatannol
Show More Show Less

RUO – Research Use Only