missing translation for 'onlineSavingsMsg'
Learn More

Piceatannol 98.0+%, TCI America™

Catalog No. P19281G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P19281G 1 g
P1928100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P19281G Supplier TCI America Supplier No. P19281G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10083-24-6
Molecular Formula C14H12O4
Molecular Weight (g/mol) 244.246
MDL Number MFCD00221715
InChI Key CDRPUGZCRXZLFL-OWOJBTEDSA-N
Synonym piceatannol, 3-hydroxyresveratol, astringinin, piceatanol, e-4-3,5-dihydroxystyryl benzene-1,2-diol, 3,3',4'5-tetrahydroxystilbene, 3,5,3',4'-tetrahydroxystilbene, 4-e-2-3,5-dihydroxyphenyl ethenyl benzene-1,2-diol, 3,3',4,5'-tetrahydroxystilbene, unii-6ks3ls0d4f
PubChem CID 667639
ChEBI CHEBI:28814
IUPAC Name 4-[(E)-2-(3,5-dihydroxyphenyl)ethenyl]benzene-1,2-diol
SMILES C1=CC(=C(C=C1C=CC2=CC(=CC(=C2)O)O)O)O

Specifications

Melting Point 235°C
Color Red-White
Boiling Point 108°C
Quantity 1 g
Formula Weight 244.25
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Piceatannol
TSCA No
Recommended Storage Refrigerator