missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Piceatannol

Catalog No. AC328452500
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC328452500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC328452500 Supplier Thermo Scientific™ Supplier No. 328452500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10083-24-6 | C14H12O4 | 244.25 g/mol

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 10083-24-6
Molecular Formula C14H12O4
Molecular Weight (g/mol) 244.25
InChI Key CDRPUGZCRXZLFL-OWOJBTEDSA-N
IUPAC Name 5-[(1E)-2-(3,4-dihydroxyphenyl)ethenyl]benzene-1,3-diol
SMILES OC1=CC(\C=C\C2=CC=C(O)C(O)=C2)=CC(O)=C1

Specifications

Melting Point 235.0°C
Quantity 250 mg
Solubility Information Solubility aq. soln.: insoluble. Other solubilities: soluble in ethanol, dmso, dimethyl formamide,, acetone, ethyl acetate, benzene
Formula Weight 244.24
Chemical Name or Material Piceatannol

RUO – Research Use Only