accessibility menu, dialog, popup

UserName

CH3OHNPOOHHOPOOHOHNHONHONHONHOHNPOOHHOPOHOOHH3CNaNaNaNa

CAS RN 1006028-37-0

IUPAC Name: {[2-(3-{3-[({3-[(5-{[2,4-bis(phosphonomethyl)phenyl]carbamoyl}-2-methylphenyl)carbamoyl]phenyl}carbamoyl)amino]benzamido}-4-methylbenzamido)-5-(phosphonomethyl)phenyl]methyl}phosphonic acid tetrasodium
Synonyms: [2-(3-{3-[({3-[(5-{[2,4-bis(phosphonomethyl)phenyl]carbamoyl}-2-methylphenyl)carbamoyl]phenyl}carbamoyl)amino]benzamido}-4-methylbenzamido)-5-(phosphonomethyl)phenyl]methylphosphonic acid tetrasodium
Molecular Weight (g/mol): 1184.78
Molecular Formula: C47H48N6Na4O17P4
InChi Key: HLUPMJLBMAPXSC-UHFFFAOYSA-N
SMILES: [Na].[Na].[Na].[Na].CC1=C(NC(=O)C2=CC(NC(=O)NC3=CC=CC(=C3)C(=O)NC3=C(C)C=CC(=C3)C(=O)NC3=C(CP(O)(O)=O)C=C(CP(O)(O)=O)C=C3)=CC=C2)C=C(C=C1)C(=O)NC1=C(CP(O)(O)=O)C=C(CP(O)(O)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results