missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ NF 546

Catalog No. 389210
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
389210 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 389210 Supplier R&D Systems™ Supplier No. 3892/10

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Tocris product, part of R&D Systems. Selective P2Y11 agonist

P2Y11 agonist (pEC50 = 6.27). Exhibits selectivity for P2Y11 over a range of P2Y and P2X receptors. Also inhibits DNA-binding activity of HMGA2 (IC50 = 5.49 μM).

Specifications

CAS 1006028-37-0
Quantity 10 mg
Product Line Tocris
Molecular Formula C47H44N6Na4O17P4
Formula Weight 1180.74
Purity >95%
Molecular Weight (g/mol) 1180.748
Chemical Name or Material NF 546
Recommended Storage Store at -20°C