accessibility menu, dialog, popup

UserName

CAS RN 100361-18-0

IUPAC Name: 7-chloro-1-cyclopropyl-6-fluoro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylic acid
Synonyms: 7-chloro-1-cyclopropyl-6-fluoro-4-oxo-1,8-naphthyridine-3-carboxylic acid
Molecular Weight (g/mol): 282.66
Molecular Formula: C12H8ClFN2O3
InChi Key: OXNZWNNMJBOZQO-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN(C2CC2)C2=NC(Cl)=C(F)C=C2C1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results