accessibility menu, dialog, popup

UserName

CAS RN 10031-52-4

IUPAC Name: dierbium(3+) octahydrate trisulfate
Synonyms: dierbium(3+) octahydrate trisulfate
Molecular Weight (g/mol): 766.81
Molecular Formula: Er2H16O20S3
InChi Key: SMNPRCXUVYGCAE-UHFFFAOYSA-H
SMILES: O.O.O.O.O.O.O.O.[Er+3].[Er+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)