missing translation for 'onlineSavingsMsg'
Learn More

Erbium(III) sulfate octahydrate, 99.9% (REO)

Catalog No. AA4009218
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4009218 50 g
AA4009206 5 g
AA4009209 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4009218 Supplier Thermo Scientific Chemicals Supplier No. 04009218

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10031-52-4

Erbium(III) sulfate octahydrate is used as a colorant in glass manufacturing and porcelain enamel glazes. High purity Erbium Sulfate is applied as dopant in making optical fiber.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10031-52-4
MDL Number MFCD00149696
Synonym erbium sulfate octahydrate, dierbium trisulfate octahydrate, erbium sulfate er2 so4 3 octahydrate, erbium sulfate hydrate er2 so4 3.8h2o, sulfuric acid, erbium 3+ salt 3:2 , octahydrate, erbium sulfate, erbium sulfate-water 2/3/8, erbium iii sulfate octahydrate reo, dierbium 3+ ion octahydrate trisulfate

Specifications

Melting Point (decomposition)
Physical Form Crystalline
Quantity 50 g
Solubility Information Soluble in water.
Formula Weight 766.82 (622.70 Anhydrous)
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Density 3.205 g/mL
Assay (REO)
Chemical Name or Material Erbium(III) sulfate octahydrate
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 236-779-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only