accessibility menu, dialog, popup

UserName

CAS RN 100-01-6

IUPAC Name: 4-nitroaniline
Synonyms: p-nitroaniline p-nitraniline 4-nitraniline
Molecular Weight (g/mol): 138.13
Molecular Formula: C6H6N2O2
InChi Key: TYMLOMAKGOJONV-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)[N+]([O-])=O
Boiling Point 
  • (3)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)